Gene Transcripts

Gene Symbol
Crh
Entrez Gene ID
12918
Gene Name
corticotropin releasing hormone
Gene Aliases
CRF, Gm1347
Chromosome Location
Chr. 3: 19693401 - 19695396 on Build GRCm38
UniGene ID
Mm.290689
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_205769.2 NP_991338.1 2 834
GenBank mRNA BC116921.1 - 1 616
BC119036.1 - 1 616 8733785

View Full Product Details