Gene Transcripts

Gene Symbol
DNAJC25
Entrez Gene ID
548645
Gene Name
DnaJ heat shock protein family (Hsp40) member C25
Gene Aliases
bA16L21.2.1
Chromosome Location
Chr. 9: 111631352 - 111654351 on Build GRCh38
UniGene ID
Hs.534196
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015882.2 NP_001015882.2 4 2061
NR_037148.1 5 2299
GenBank mRNA AF447177.1 - 5 2268
AK095452.1 - 5 2282

View Full Product Details