Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009662.2 | NP_033792.1 | 8 | 1245 | |
GenBank mRNA | AK089344.1 | - | 8 | 1199 | |
AK136868.1 | - | 8 | 1214 | ||
AK137481.1 | - | 8 | 1203 | ||
AK169938.1 | - | 8 | 1202 | ||
AK170016.1 | - | 8 | 1102 | ||
AK170434.1 | - | 8 | 1202 | ||
AK170879.1 | - | 8 | 1201 | ||
AK171191.1 | - | 8 | 1194 | ||
AK171413.1 | - | 8 | 1202 | ||
BC139102.1 | - | 8 | 1172 | 8862124 | |
BC141213.1 | - | 8 | 1172 | 9055825 | |
L42198.1 | - | 8 | 1091 |