Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012148.1 | NP_001012148.1 | 15 | 3364 | |
XM_006249461.1 | XP_006249523.1 | 15 | 3904 | ||
XM_006249462.2 | XP_006249524.1 | 15 | 3248 | ||
GenBank mRNA | BC085774.1 | - | 15 | 3364 |