Gene Transcripts

Gene Symbol
Ccnl2
Entrez Gene ID
298686
Gene Name
cyclin L2
Gene Aliases
Chromosome Location
Chr. 5: 173256308 - 173268279 on Build Rnor_6.0
UniGene ID
Rn.232195
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013094.1 NP_001013112.1 6 1034
GenBank mRNA BC088316.1 - 6 1034
FQ211043.1 - 6 1095

View Full Product Details