Gene Transcripts

Gene Symbol
Dusp18
Entrez Gene ID
305477
Gene Name
dual specificity phosphatase 18
Gene Aliases
Chromosome Location
Chr. 14: 84150484 - 84168173 on Build Rnor_6.0
UniGene ID
Rn.19584
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013128.2 NP_001013146.2 4 940
XM_006251299.2 XP_006251361.1 3 1986
GenBank mRNA BC079285.1 - 4 922

View Full Product Details