Gene Transcripts

Gene Symbol
Sccpdh
Entrez Gene ID
305021
Gene Name
saccharopine dehydrogenase (putative)
Gene Aliases
RGD1311440
Chromosome Location
Chr. 13: 97941720 - 97963474 on Build Rnor_6.0
UniGene ID
Rn.23989
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013985.1 NP_001014007.1 4 595
GenBank mRNA BC079215.1 - 4 595

Gene Symbol
LOC103689999
Entrez Gene ID
103689999
Gene Name
saccharopine dehydrogenase-like oxidoreductase
Gene Aliases
Chromosome Location
Chr. 13: 97701878 - 97724165 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769879.1 XP_008768101.1 4 777

View Full Product Details