Gene Transcripts

Gene Symbol
RBM28
Entrez Gene ID
55131
Gene Name
RNA binding motif protein 28
Gene Aliases
ANES
Chromosome Location
Chr. 7: 128304120 - 128343915 on Build GRCh38
UniGene ID
Hs.274263
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018077.2 NP_060547.2 1, 2 241
XM_017012390.1 XP_016867879.1 1, 2 241
GenBank mRNA AK001239.1 - 1, 2 241
BC013889.2 - 1, 2 195 3926178
DA566130.1 - 1, 2 241
KU178685.1 - 1, 2 126
KU178686.1 - 1, 2 126

View Full Product Details