Gene Transcripts

Gene Symbol
CT83
Entrez Gene ID
203413
Gene Name
cancer/testis antigen 83
Gene Aliases
CXorf61, KK-LC-1, KKLC1
Chromosome Location
Chr. X: 116461686 - 116463033 on Build GRCh38
UniGene ID
Hs.97892
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017978.3 NP_001017978.1 2 337
GenBank mRNA BC062223.1 - 2 284

View Full Product Details