Gene Transcripts

Gene Symbol
Rhox9
Entrez Gene ID
298352
Gene Name
reproductive homeobox 9
Gene Aliases
Chromosome Location
Chr. Un|NW_007906942: 9496 - 10900 on Build Rnor_6.0
UniGene ID
Rn.144519
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024874.1 NP_001020045.1 4 814
GenBank mRNA DQ058656.1 - 4 814

View Full Product Details