Gene Transcripts

Gene Symbol
KU-MEL-3
Entrez Gene ID
497048
Gene Name
uncharacterized LOC497048
Gene Aliases
Chromosome Location
Chr. 6: 4610565 - 4611919 on Build GRCh38
UniGene ID
Hs.591355
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_122107.1 2 424
GenBank mRNA AY870321.1 - 2 377
BC130651.1 - 2 272 40146682
BC130655.1 - 2 277 40146686

View Full Product Details