Gene Transcripts

Gene Symbol
Ier5
Entrez Gene ID
498256
Gene Name
immediate early response 5
Gene Aliases
Chromosome Location
Chr. 13: 72742210 - 72744302 on Build Rnor_6.0
UniGene ID
Rn.8590
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025137.2 NP_001020308.2 1 1315
GenBank mRNA BC091181.1 - 2 591
CO571883.1 - 2 590

View Full Product Details