Gene Transcripts

Gene Symbol
Adam5
Entrez Gene ID
498654
Gene Name
a disintegrin and metallopeptidase domain 5
Gene Aliases
tMDCII
Chromosome Location
Chr. 16: 72010051 - 72085655 on Build Rnor_6.0
UniGene ID
Rn.92321
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020303.1 NP_064699.1 18 2097
XM_006253321.2 XP_006253383.1 18 2147
XM_006253322.2 XP_006253384.1 18 2084
GenBank mRNA BC079016.1 - 18 2067
BC091169.1 - 18 2097 7112347

View Full Product Details