Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028478.2 | NP_082754.2 | 11 | 1513 | |
GenBank mRNA | AK005472.1 | - | 11 | 1461 | |
AK132288.1 | - | 11 | 1475 | ||
BC031808.1 | - | 11 | 1485 | ||
BC039934.1 | - | 11 | 1560 | ||
BC075725.1 | - | 8 | 2851 |