Gene Transcripts

Gene Symbol
Lrrc2
Entrez Gene ID
74249
Gene Name
leucine rich repeat containing 2
Gene Aliases
2400002D05Rik, 4933431K03Rik
Chromosome Location
Chr. 9: 110951545 - 110984066 on Build GRCm38
UniGene ID
Mm.307615
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028838.2 NP_083114.2 5 693
GenBank mRNA AJ428068.1 - 5 692
AK010252.1 - 5 692

View Full Product Details