Gene Transcripts

Gene Symbol
Gstm1
Entrez Gene ID
14862
Gene Name
glutathione S-transferase, mu 1
Gene Aliases
Gstb-1, Gstb1
Chromosome Location
Chr. 3: 108012250 - 108017973 on Build GRCm38
UniGene ID
Mm.37199
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010358.5 NP_034488.1 8 825
GenBank mRNA BC003822.1 - 8 648
BC046758.1 - 8 678 5693552
BC091763.1 - 8 658 5102093
J03952.1 - 8 639
J04632.1 - 8 826

View Full Product Details