Gene Transcripts

Gene Symbol
LGALS16
Entrez Gene ID
148003
Gene Name
galectin 16
Gene Aliases
Chromosome Location
Chr. 19: 39655894 - 39660647 on Build GRCh38
UniGene ID
Hs.667238
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190441.2 NP_001177370.2 3 301
GenBank mRNA BX095861.1 - 3 261
FJ613352.1 - 3 301

View Full Product Details