Gene Transcripts

Gene Symbol
RBM41
Entrez Gene ID
55285
Gene Name
RNA binding motif protein 41
Gene Aliases
Chromosome Location
Chr. X: 107061885 - 107118827 on Build GRCh38
UniGene ID
Hs.139053
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171080.1 NP_001164551.1 3 245
NM_001324242.1 NP_001311171.1 3 245
NM_001324243.1 NP_001311172.1 3 338
NM_001324244.1 NP_001311173.1 3 245
NM_018301.4 NP_060771.3 3 245
NR_136728.1 3 245
NR_136729.1 4 339
NR_136730.1 3 245
NR_136731.1 3 338
NR_136732.1 3 338
XM_011530980.2 XP_011529282.1 3 337
XM_011530981.2 XP_011529283.1 3 337
GenBank mRNA AK001878.1 - 3 218
BC006986.1 - 3 222 3995903
DA255574.1 - 3 245
DB132398.1 - 3 245

View Full Product Details