Gene Transcripts

Gene Symbol
SNTG1
Entrez Gene ID
54212
Gene Name
syntrophin gamma 1
Gene Aliases
G1SYN, SYN4
Chromosome Location
Chr. 8: 49909726 - 50796656 on Build GRCh38
UniGene ID
Hs.584914
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287813.2 NP_001274742.1 1 147
NM_001321775.1 NP_001308704.1 1 147
GenBank mRNA AK314084.1 - 1 84
HY017401.1 - 1 148

View Full Product Details