Gene Transcripts

Gene Symbol
HEATR5B
Entrez Gene ID
54497
Gene Name
HEAT repeat containing 5B
Gene Aliases
Chromosome Location
Chr. 2: 36980999 - 37084384 on Build GRCh38
UniGene ID
Hs.744084
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011532934.2 XP_011531236.1 1 466
XM_017004379.1 XP_016859868.1 1 221

View Full Product Details