Gene Transcripts
- Gene Symbol
- FDPSP2
- Entrez Gene ID
- 619190
- Gene Name
- farnesyl diphosphate synthase pseudogene 2
- Gene Aliases
- FDPSL2, FDPSL2A, FPSL2
- Chromosome Location
- Chr. 7: 76470020 - 76475000 on Build GRCh38
- UniGene ID
- Hs.609978
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003262.1 |
|
1 |
4420 |
|
GenBank mRNA |
BC050292.1 |
- |
2 |
3144 |
|
View Full Product Details