Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320041.1 | NP_001306970.1 | 3 | 323 | |
NM_018456.5 | NP_060926.2 | 4 | 460 | ||
XM_005247618.3 | XP_005247675.1 | 5 | 610 | ||
XM_017006861.1 | XP_016862350.1 | 5 | 332 | ||
XM_017006862.1 | XP_016862351.1 | 3 | 232 | ||
XM_017006863.1 | XP_016862352.1 | 4 | 403 | ||
GenBank mRNA | AF517829.1 | - | 4 | 405 | |
AK301560.1 | - | 4 | 403 | ||
AY049020.1 | - | 4 | 425 | ||
BC014209.2 | - | 4 | 420 | 4765823 | |
BU941228.1 | - | 456 | |||
HY005912.1 | - | 4 | 461 |