Gene Transcripts

Gene Symbol
PVT1
Entrez Gene ID
5820
Gene Name
Pvt1 oncogene (non-protein coding)
Gene Aliases
LINC00079, MYC, NCRNA00079, onco-lncRNA-100
Chromosome Location
Chr. 8: 127794533 - 128101253 on Build GRCh38
UniGene ID
Hs.133107
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003367.3 7 1419
GenBank mRNA BC041065.1 - 6 1178
BX648921.1 - 1 533
D79458.1 - 4 418

View Full Product Details