Gene Transcripts

Gene Symbol
BEX4
Entrez Gene ID
56271
Gene Name
brain expressed X-linked 4
Gene Aliases
BEXL1
Chromosome Location
Chr. X: 103215092 - 103217200 on Build GRCh38
UniGene ID
Hs.184736
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080425.3 NP_001073894.1 3 488
NM_001127688.2 NP_001121160.1 2 405
GenBank mRNA AK000959.1 - 3 443
AL544124.3 - 2 381
AV726729.1 - 3 488
AY833563.1 - 3 443

View Full Product Details