Gene Transcripts

Gene Symbol
UNK
Entrez Gene ID
85451
Gene Name
unkempt family zinc finger
Gene Aliases
UNKEMPT, ZC3H5, ZC3HDC5
Chromosome Location
Chr. 17: 75784771 - 75825805 on Build GRCh38
UniGene ID
Hs.584768
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017025248.1 XP_016880737.1 1 65
XM_017025250.1 XP_016880739.1 1 65

View Full Product Details