Gene Transcripts
- Gene Symbol
- TANC2
- Entrez Gene ID
- 26115
- Gene Name
- tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2
- Gene Aliases
- ROLSA, rols
- Chromosome Location
- Chr. 17: 62966523 - 63427706 on Build GRCh38
- UniGene ID
- Hs.410889
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025185.3 |
NP_079461.2
|
4, 5 |
379 |
|
XM_005257203.4 |
XP_005257260.1
|
5, 6 |
606 |
|
XM_006721806.3 |
XP_006721869.1
|
6, 7 |
828 |
|
XM_006721807.3 |
XP_006721870.1
|
6, 7 |
828 |
|
XM_006721810.3 |
XP_006721873.1
|
6, 7 |
828 |
|
XM_006721811.3 |
XP_006721874.1
|
6, 7 |
828 |
|
XM_011524597.2 |
XP_011522899.1
|
6, 7 |
828 |
|
XM_011524598.2 |
XP_011522900.1
|
6, 7 |
828 |
|
XM_011524599.1 |
XP_011522901.1
|
6, 7 |
654 |
|
XM_011524600.2 |
XP_011522902.1
|
5, 6 |
717 |
|
XM_011524601.2 |
XP_011522903.1
|
6, 7 |
717 |
|
XM_011524602.2 |
XP_011522904.1
|
5, 6 |
606 |
|
XM_017024429.1 |
XP_016879918.1
|
6, 7 |
654 |
|
XM_017024430.1 |
XP_016879919.1
|
2, 3 |
3995 |
|
XM_017024431.1 |
XP_016879920.1
|
6, 7 |
828 |
|
XM_017024432.1 |
XP_016879921.1
|
6, 7 |
828 |
|
View Full Product Details