Gene Transcripts

Gene Symbol
PLEKHM2
Entrez Gene ID
23207
Gene Name
pleckstrin homology and RUN domain containing M2
Gene Aliases
SKIP
Chromosome Location
Chr. 1: 15681506 - 15734769 on Build GRCh38
UniGene ID
Hs.646775
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015164.2 NP_055979.2 12 2188
XM_005245790.3 XP_005245847.1 11 2323
XM_005245791.4 XP_005245848.1 7 1456
XM_017000757.1 XP_016856246.1 12 2095
XM_017000758.1 XP_016856247.1 11 2035
GenBank mRNA AB020649.1 - 12 1964
BC008002.1 - 4 225
BC016488.1 - 4 870
BC030545.1 - 3 168
BC040441.1 - 12 2191
BC042103.1 - 4 518
BC068599.1 - 4 543

View Full Product Details