Gene Transcripts

Gene Symbol
BAK1
Entrez Gene ID
578
Gene Name
BCL2 antagonist/killer 1
Gene Aliases
BAK, BAK-LIKE, BCL2L7, CDN1
Chromosome Location
Chr. 6: 33572546 - 33580293 on Build GRCh38
UniGene ID
Hs.485139
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001188.3 NP_001179.1 6 1390
XM_011514779.2 XP_011513081.1 7 1501
XM_011514780.1 XP_011513082.1 6 1311
GenBank mRNA AK091807.1 - 6 1255
AK293547.1 - 6 1320
BC004431.1 - 1 1301 2819507
BC110337.1 - 7 1320 5755200
CR627020.1 - 1 6724
U16811.1 - 6 1283
U23765.1 - 6 1153
X84213.1 - 6 1279

View Full Product Details