Gene Transcripts

Gene Symbol
BAIAP2L1
Entrez Gene ID
55971
Gene Name
BAI1 associated protein 2 like 1
Gene Aliases
IRTKS
Chromosome Location
Chr. 7: 98291650 - 98401115 on Build GRCh38
UniGene ID
Hs.656063
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018842.4 NP_061330.2 3 413
GenBank mRNA AF119666.2 - 3 366
BC013888.2 - 3 366 3842949
BI560834.1 - 3 416
DA442866.1 - 3 402

View Full Product Details