Gene Transcripts

Gene Symbol
DDIT4
Entrez Gene ID
54541
Gene Name
DNA damage inducible transcript 4
Gene Aliases
Dig2, REDD-1, REDD1
Chromosome Location
Chr. 10: 72273919 - 72276039 on Build GRCh38
UniGene ID
Hs.744875
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019058.2 NP_061931.1 3 588
GenBank mRNA AF335324.1 - 3 604
AK000507.1 - 3 583
AL136668.1 - 3 587
AY090097.1 - 3 504
BC000708.1 - 3 494 3507244
BC007714.1 - 3 576 4302878
BC015236.1 - 3 562 3906374

View Full Product Details