Gene Transcripts

Gene Symbol
C16orf91
Entrez Gene ID
283951
Gene Name
chromosome 16 open reading frame 91
Gene Aliases
CCSMST1, URLC5, gs103
Chromosome Location
Chr. 16: 1419744 - 1420800 on Build GRCh38
UniGene ID
Hs.58373
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001272051.1 NP_001258980.1 2 735
GenBank mRNA AB101207.1 - 2 679
BC092497.1 - 2 679

View Full Product Details