Gene Transcripts

Gene Symbol
SPRR2D
Entrez Gene ID
6703
Gene Name
small proline rich protein 2D
Gene Aliases
Chromosome Location
Chr. 1: 153039725 - 153041144 on Build GRCh38
UniGene ID
Hs.505327
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006945.4 NP_008876.3 2 485
XM_017002175.1 XP_016857664.1 2 517
GenBank mRNA AK291317.1 - 2 487
M21302.1 - 2 485

View Full Product Details