Gene Transcripts

Gene Symbol
CFAP221
Entrez Gene ID
200373
Gene Name
cilia and flagella associated protein 221
Gene Aliases
FAP221, PCDP1
Chromosome Location
Chr. 2: 119544392 - 119688610 on Build GRCh38
UniGene ID
Hs.496753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271049.1 NP_001257978.1 14, 15 1539
NR_073132.1 15, 16 1684
NR_073133.1 13, 14 1438
XM_006712353.3 XP_006712416.1 14, 15 1579
XM_017003550.1 XP_016859039.1 14, 15 1579
XM_017003551.1 XP_016859040.1 14, 15 1706
XM_017003552.1 XP_016859041.1 14, 15 1592
XM_017003553.1 XP_016859042.1 14, 15 1925
XM_017003554.1 XP_016859043.1 13, 14 1421
XM_017003555.1 XP_016859044.1 13, 14 1421
XM_017003556.1 XP_016859045.1 13, 14 1424
XM_017003557.1 XP_016859046.1 12, 13 1354
XM_017003558.1 XP_016859047.1 14, 15 1579
XM_017003559.1 XP_016859048.1 11, 12 2550
GenBank mRNA AK126306.1 - 1, 2 89
AK293665.1 - 13, 14 1421
BC036530.1 - 15, 16 1650 4828213

View Full Product Details