Gene Transcripts

Gene Symbol
INSC
Entrez Gene ID
387755
Gene Name
inscuteable homolog (Drosophila)
Gene Aliases
Chromosome Location
Chr. 11: 15111416 - 15269675 on Build GRCh38
UniGene ID
Hs.591997
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031853.4 NP_001027024.3 9 1318
NM_001042536.2 NP_001036001.1 9 1242
NM_001278313.1 NP_001265242.1 8 1204
NM_001278314.1 NP_001265243.1 9 1309
NM_001278315.1 NP_001265244.1 8 1204
NM_001278316.1 NP_001265245.1 7 1078
XM_006718227.3 XP_006718290.1 10 1371
XM_011520085.1 XP_011518387.1 9 1246
XM_011520086.1 XP_011518388.1 9 1246
XM_017017697.1 XP_016873186.1 10 1283
XM_017017698.1 XP_016873187.1 9 1246
GenBank mRNA AB231745.1 - 8 1198
AB231746.1 - 9 1303
AB231747.1 - 7 1072
AB231748.1 - 4 573
AB236158.1 - 9 1242
AB236159.1 - 9 1318
BC127700.1 - 8 1204 40127962

View Full Product Details