Gene Transcripts

Gene Symbol
SAMD5
Entrez Gene ID
389432
Gene Name
sterile alpha motif domain containing 5
Gene Aliases
dJ875H10.1
Chromosome Location
Chr. 6: 147508692 - 147948291 on Build GRCh38
UniGene ID
Hs.567973
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030060.2 NP_001025231.1 2 1137
GenBank mRNA BX642659.1 - 2 503

View Full Product Details