Gene Transcripts

Gene Symbol
RGL3
Entrez Gene ID
57139
Gene Name
ral guanine nucleotide dissociation stimulator like 3
Gene Aliases
Chromosome Location
Chr. 19: 11394056 - 11419342 on Build GRCh38
UniGene ID
Hs.375142
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035223.3 NP_001030300.3 5 642
NM_001161616.2 NP_001155088.2 5 642
GenBank mRNA AK057147.1 - 5 605
AK298281.1 - 4 782
BC014426.1 - 5 630
BC101756.1 - 5 628 8069262
BC111958.1 - 5 628 8327426
BC143530.1 - 5 628 9052039

View Full Product Details