Gene Transcripts
- Gene Symbol
- Rps10
- Entrez Gene ID
- 81773
- Gene Name
- ribosomal protein S10
- Gene Aliases
- Chromosome Location
- Chr. 20: 7215762 - 7220331 on Build Rnor_6.0
- UniGene ID
- Rn.3359
- Species
- Rattus norvegicus
- Gene Symbol
- Rpl30
- Entrez Gene ID
- 64640
- Gene Name
- ribosomal protein L30
- Gene Aliases
- Chromosome Location
- Chr. 7: 73213538 - 73216431 on Build Rnor_6.0
- UniGene ID
- Rn.161950
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
FQ229603.1 |
- |
|
260 |
|
- Gene Symbol
- Rps10l1
- Entrez Gene ID
- 100363439
- Gene Name
- ribosomal protein S10-like 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 75429715 - 75430282 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details