Gene Transcripts

Gene Symbol
Stx11
Entrez Gene ID
292483
Gene Name
syntaxin 11
Gene Aliases
Chromosome Location
Chr. 1: 7039160 - 7064870 on Build Rnor_6.0
UniGene ID
Rn.200930
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025638.2 NP_001020809.2 2 542
XM_008758601.1 XP_008756823.1 2 590
GenBank mRNA BC099159.1 - 1 209
CK478859.1 - 2 542

View Full Product Details