Gene Transcripts

Gene Symbol
Ubxn1
Entrez Gene ID
293719
Gene Name
UBX domain protein 1
Gene Aliases
2B28, RGD1309471
Chromosome Location
Chr. 1: 225067909 - 225071946 on Build Rnor_6.0
UniGene ID
Rn.198539
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034829.1 NP_001030001.1 4 280
XM_006230975.1 XP_006231037.1 5 457
GenBank mRNA AJ277751.1 - 4 280

View Full Product Details