Gene Transcripts

Gene Symbol
Gpr35
Entrez Gene ID
367315
Gene Name
G protein-coupled receptor 35
Gene Aliases
Chromosome Location
Chr. 9: 100129496 - 100140673 on Build Rnor_6.0
UniGene ID
Rn.154784
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037359.1 NP_001032436.1 1 493
XM_006245537.2 XP_006245599.1 2 636
XM_006245539.2 XP_006245601.1 2 851
XM_006245540.2 XP_006245602.1 2 848
GenBank mRNA AB240684.1 - 1 493

View Full Product Details