Gene Transcripts
- Gene Symbol
- Rhox4e
- Entrez Gene ID
- 194856
- Gene Name
- reproductive homeobox 4E
- Gene Aliases
- Rhox4, Rhox4.5, Rhox4d
- Chromosome Location
- Chr. X: 37557412 - 37562282 on Build GRCm38
- UniGene ID
- Mm.379498
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC094891.1 |
- |
3, 4 |
701 |
|
View Full Product Details