Gene Transcripts

Gene Symbol
Fhdc1
Entrez Gene ID
229474
Gene Name
FH2 domain containing 1
Gene Aliases
6330505N24Rik, Gm126
Chromosome Location
Chr. 3: 84442191 - 84480439 on Build GRCm38
UniGene ID
Mm.331107
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033301.4 NP_001028473.2 11, 12 1595
NM_001205355.1 NP_001192284.1 11, 12 1666
GenBank mRNA AK031946.1 - 10, 11 1068
AK145217.1 - 11, 12 1586
AK173244.1 - 11, 12 1666
BC137632.1 - 11, 12 1635 8860651

View Full Product Details