Gene Transcripts

Gene Symbol
Dhrsx
Entrez Gene ID
236082
Gene Name
dehydrogenase/reductase (SDR family) X chromosome
Gene Aliases
C80877, D4Ertd296e, Dhrsxy, Pscad
Chromosome Location
Chr. 4|NT_166291: 15800 - 36213 on Build GRCm38
UniGene ID
Mm.305345
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033326.2 NP_001028498.2 7 1808
GenBank mRNA AK030649.1 - 6 1736
AK157461.1 - 7 1493
BC138597.1 - 7 1450 8861617
BC138599.1 - 7 1450 8861619

View Full Product Details