Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020358.2 | NP_065091.1 | 8 | 1780 | |
XM_017018027.1 | XP_016873516.1 | 5 | 1224 | ||
GenBank mRNA | AB037682.1 | - | 8 | 1780 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195234.1 | NP_001182163.1 | 8 | 1780 | |
XM_017018126.1 | XP_016873615.1 | 5 | 1224 |