Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014787.1 | NP_001014787.1 | 8 | 2009 | |
XM_006250286.2 | XP_006250348.1 | 8 | 2168 | ||
XM_006250288.2 | XP_006250350.1 | 7 | 2110 | ||
XM_006250289.2 | XP_006250351.1 | 7 | 1947 | ||
GenBank mRNA | BC092192.1 | - | 8 | 2009 |