Gene Transcripts
- Gene Symbol
- YIF1A
- Entrez Gene ID
- 10897
- Gene Name
- Yip1 interacting factor homolog A, membrane trafficking protein
- Gene Aliases
- 54TM, FinGER7, YIF1, YIF1P
- Chromosome Location
- Chr. 11: 66284580 - 66289167 on Build GRCh38
- UniGene ID
- Hs.446445
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF004876.1 |
- |
5 |
555 |
|
View Full Product Details