Gene Transcripts

Gene Symbol
Nova2
Entrez Gene ID
384569
Gene Name
neuro-oncological ventral antigen 2
Gene Aliases
Gm1424
Chromosome Location
Chr. 7: 18925888 - 18965319 on Build GRCm38
UniGene ID
Mm.262724
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029877.3 NP_001025048.2 3 490
GenBank mRNA CJ126972.1 - 3 449

View Full Product Details