Gene Transcripts

Gene Symbol
Gdf7
Entrez Gene ID
238057
Gene Name
growth differentiation factor 7
Gene Aliases
BMP12
Chromosome Location
Chr. 12: 8291018 - 8302088 on Build GRCm38
UniGene ID
Mm.271308
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013527.1 NP_038555.1 2 1103
GenBank mRNA AF525752.1 - 2 1103

View Full Product Details