Gene Transcripts

Gene Symbol
Mettl7a1
Entrez Gene ID
70152
Gene Name
methyltransferase like 7A1
Gene Aliases
2210414H16Rik, 3300001H21Rik, Aam-B, Mettl7a, UbiE1
Chromosome Location
Chr. 15: 100304145 - 100328665 on Build GRCm38
UniGene ID
Mm.488086
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027334.3 NP_081610.2 2 614
GenBank mRNA AB253943.1 - 2 585
AJ536232.1 - 2 588
AK076169.1 - 2 616
BC033446.1 - 2 591
BC047149.1 - 2 598
BC055034.1 - 2 605 6306873
BC060144.1 - 2 593
BC171988.1 - 2 608 9054274
BC171989.1 - 2 608 9054275

View Full Product Details