Gene Transcripts

Gene Symbol
Fpr-rs6
Entrez Gene ID
321020
Gene Name
formyl peptide receptor, related sequence 6
Gene Aliases
Chromosome Location
Chr. 17: 20182078 - 20183097 on Build GRCm38
UniGene ID
Mm.377882
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177316.2 NP_796290.2 1 616
GenBank mRNA BC104318.1 - 1 616
BC104319.1 - 1 616 40044732

View Full Product Details